Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000365.5 | NP_000356.1 | 7 | 729 | ||
NM_001159287.1 | NP_001152759.1 | 7 | 838 | |||
NM_001258026.1 | NP_001244955.1 | 7 | 972 | |||
GenBank mRNA | AK129488.1 | - | 7 | 741 | ||
AK222638.1 | - | 7 | 718 | |||
AK298809.1 | - | 7 | 973 | |||
AK300661.1 | - | 7 | 617 | |||
AK313282.1 | - | 7 | 725 | |||
BC007086.1 | - | 7 | 725 | 4244564 | ||
BC007812.2 | - | 7 | 710 | 4298901 | ||
BC009329.2 | - | 7 | 716 | 4126160 | ||
BC011611.2 | - | 7 | 715 | 3956298 | ||
BC015100.1 | - | 7 | 730 | 3829453 | ||
BC017165.1 | - | 6 | 475 | |||
BC017917.1 | - | 7 | 721 | 4296431 | ||
BC018899.1 | - | 1140 | ||||
CR541702.1 | - | 7 | 691 | |||
EU794668.1 | - | 7 | 730 | |||
M10036.1 | - | 7 | 1058 |