Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM227A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013647.1 | NP_001013669.1 | 6 | 1062 | ||
NM_001291030.1 | NP_001277959.1 | 6 | 1102 | |||
XM_006724291.2 | XP_006724354.2 | 6 | 1062 | |||
XM_006724292.2 | XP_006724355.2 | 6 | 1062 | |||
XM_006724293.2 | XP_006724356.1 | 6 | 1062 | |||
XM_006724294.2 | XP_006724357.2 | 6 | 1102 | |||
XM_006724295.2 | XP_006724358.2 | 6 | 1102 | |||
XM_011530308.2 | XP_011528610.1 | 6 | 767 | |||
XM_011530309.2 | XP_011528611.1 | 6 | 1062 | |||
XM_011530310.1 | XP_011528612.1 | 6 | 1062 | |||
XM_011530311.1 | XP_011528613.1 | 5 | 939 | |||
XM_011530312.1 | XP_011528614.1 | 4 | 339 | |||
XM_011530313.2 | XP_011528615.1 | 4 | 291 | |||
XM_011530314.1 | XP_011528616.1 | 5 | 992 | |||
GenBank mRNA | AK301819.1 | - | 6 | 776 |