Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011532697.2 | XP_011530999.1 | 1 | 5531 | ||
GenBank mRNA | BC037242.1 | - | 1 | 378 |