Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM212A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203370.1 | NP_976248.1 | 2 | 746 | ||
XM_017006372.1 | XP_016861861.1 | 2 | 790 | |||
GenBank mRNA | BC012170.1 | - | 2 | 746 | 4642270 |