Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM153C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038353.1 | 5 | 267 | |||
GenBank mRNA | AK294762.1 | - | 5 | 261 | ||
BC101338.1 | - | 4 | 301 | 40025116 | ||
BC101339.2 | - | 6 | 526 | |||
BC101340.2 | - | 4 | 301 | |||
BC101341.2 | - | 4 | 301 | 40025123 | ||
BC131737.1 | - | 4 | 301 | 40025113 | ||
BC146883.1 | - | 4 | 513 | 9056811 | ||
BC146888.1 | - | 4 | 513 | 9056816 | ||
BC171810.1 | - | 4 | 514 | 9054464 | ||
DB030341.1 | - | 5 | 268 |
Gene Symbol: FAM153B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265615.1 | NP_001252544.1 | 4 | 172 | ||
GenBank mRNA | BC028606.1 | - | 5 | 413 | 5266126 |