Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195032.1 | NP_001181961.1 | 2 | 181 | ||
NM_001302839.1 | NP_001289768.1 | 2 | 515 | |||
NM_001302840.1 | NP_001289769.1 | 3 | 276 | |||
NM_001302841.1 | NP_001289770.1 | 2 | 175 | |||
XM_005266788.4 | XP_005266845.1 | 2 | 522 | |||
XM_011535374.2 | XP_011533676.1 | 2 | 522 | |||
XM_017010142.1 | XP_016865631.1 | 2 | 112 | |||
XM_017010143.1 | XP_016865632.1 | 2 | 189 | |||
XM_017010144.1 | XP_016865633.1 | 2 | 184 | |||
XM_017010145.1 | XP_016865634.1 | 1 | 87 | |||
XM_017010146.1 | XP_016865635.1 | 1 | 82 | |||
XM_017010147.1 | XP_016865636.1 | 2 | 172 | |||
GenBank mRNA | BF688381.1 | - | 2 | 182 | ||
HY052980.1 | - | 3 | 277 | |||
HY205185.1 | - | 2 | 371 | |||
W37337.1 | - | 2 | 103 |