Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136205.2 | NP_001129677.1 | 3 | 773 | ||
NM_001142730.2 | NP_001136202.1 | 3 | 2037 | |||
NM_001258221.1 | NP_001245150.1 | 3 | 314 | |||
NM_001258222.1 | NP_001245151.1 | 3 | 270 | |||
NM_198991.3 | NP_945342.1 | 4 | 353 | |||
XM_017025708.1 | XP_016881197.1 | 3 | 563 | |||
GenBank mRNA | AF542549.1 | - | 4 | 360 | ||
AK289870.1 | - | 3 | 459 | |||
BC063652.1 | - | 3 | 494 | |||
BI667790.1 | - | 4 | 322 | |||
BI915955.1 | - | 4 | 266 | |||
BQ129398.1 | - | 3 | 300 | |||
DB285442.1 | - | 3 | 271 | |||
DN994590.1 | - | 3 | 308 |