Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100505478 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199233.1 | NP_001186162.1 | 4 | 307 | ||
NR_133056.1 | 2 | 76 | ||||
GenBank mRNA | DB452574.1 | - | 4 | 269 | ||
DB456645.1 | - | 4 | 261 | |||
HY015952.1 | - | 2 | 59 |