Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018837.1 | NP_001018238.1 | 3 | 456 | ||
NM_006118.3 | NP_006109.2 | 3 | 600 | |||
GenBank mRNA | AK290626.1 | - | 3 | 522 | ||
AK294298.1 | - | 2 | 701 | |||
AK300676.1 | - | 3 | 578 | |||
AK310000.1 | - | 3 | 443 | |||
AK313291.1 | - | 3 | 531 | |||
BC005240.1 | - | 3 | 652 | 3931805 | ||
BC014314.1 | - | 3 | 521 | 3931852 | ||
BC015209.1 | - | 3 | 493 | 3883802 | ||
BF668367.1 | - | 430 | ||||
EU190982.1 | - | 2 | 709 | |||
EU190983.1 | - | 3 | 517 | |||
KU178523.1 | - | 3 | 439 | |||
KU178524.1 | - | 3 | 295 | |||
U68566.1 | - | 3 | 600 |