Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC390937 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017027573.1 | XP_016883062.1 | 5 | 1140 | ||
GenBank mRNA | DR731221.1 | - | 2 | 288 |