Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032186.4 | NP_115562.3 | 1 | 33 | ||
NM_201269.2 | NP_958357.1 | 1 | 33 | |||
XM_005271260.4 | XP_005271317.1 | 1 | 45 | |||
XM_011542258.2 | XP_011540560.1 | 1 | 47 | |||
XM_011542259.2 | XP_011540561.1 | 1 | 45 | |||
XM_017002487.1 | XP_016857976.1 | 1 | 45 | |||
XM_017002489.1 | XP_016857978.1 | 1 | 43 | |||
XM_017002490.1 | XP_016857979.1 | 1 | 43 | |||
XM_017002494.1 | XP_016857983.1 | 1 | 33 | |||
GenBank mRNA | DB001551.1 | - | 1 | 142 |