Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100132057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111930.1 | 8 | 989 | |||
GenBank mRNA | BX464027.2 | - | 1 | 740 | ||
DB343069.1 | - | 2 | 493 |