Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003093.2 | NP_003084.1 | 4 | 622 | ||
NR_029472.1 | 3 | 579 | ||||
GenBank mRNA | AI356808.1 | - | 3 | 591 | ||
AK223098.1 | - | 4 | 196 | |||
AK311955.1 | - | 4 | 202 | |||
BC121082.1 | - | 4 | 191 | 40119776 | ||
BC121083.1 | - | 4 | 191 | 40119777 | ||
BU153661.1 | - | 3 | 187 | |||
BU508721.1 | - | 3 | 148 | |||
M18465.1 | - | 2 | 100 | |||
X12517.1 | - | 4 | 187 |