Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000828.4 | NP_000819.3 | 16 | 4287 | ||
NM_007325.4 | NP_015564.4 | 16 | 4287 | |||
GenBank mRNA | BY799542.2 | - | 1 | 573 | ||
DB505790.1 | - | 1 | 30 |