Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMTOR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008395.3 | NP_001008396.1 | 4 | 300 | ||
NM_001318236.1 | NP_001305165.1 | 4 | 317 | |||
NM_001318237.1 | NP_001305166.1 | 3 | 219 | |||
NR_134526.1 | 4 | 286 | ||||
NR_134527.1 | 5 | 342 | ||||
NR_134528.1 | 4 | 299 | ||||
NR_134529.1 | 4 | 279 | ||||
NR_134530.1 | 4 | 316 | ||||
XM_017012198.1 | XP_016867687.1 | 6 | 687 | |||
XM_017012199.1 | XP_016867688.1 | 6 | 704 | |||
GenBank mRNA | AA772120.1 | - | 1 | 335 | ||
AJ714519.1 | - | 4 | 280 | |||
BC063401.1 | - | 4 | 280 | 5194116 | ||
BC105609.1 | - | 4 | 290 | 30321610 | ||
BC130553.1 | - | 4 | 247 | 40146584 | ||
BC130559.1 | - | 4 | 247 | 40146590 | ||
BF978089.1 | - | 299 | ||||
BG706819.1 | - | 4 | 300 | |||
BG715626.1 | - | 4 | 296 | |||
BG819624.1 | - | 4 | 260 | |||
BI598677.1 | - | 4 | 303 | |||
BI771662.1 | - | 3 | 205 | |||
CU450474.1 | - | 4 | 353 | |||
DB498044.1 | - | 4 | 295 | |||
DB539334.2 | - | 3 | 222 |