Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024497.2 | 8 | 5129 | |||
GenBank mRNA | AK128746.1 | - | 2 | 3588 | ||
BQ000710.1 | - | 1 | 138 |