Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM180B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164379.1 | NP_001157851.1 | 1 | 105 | ||
GenBank mRNA | BC065704.1 | - | 1 | 90 | ||
CB999847.1 | - | 1 | 108 |