Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR100HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024430.2 | 4 | 2735 | |||
NR_137175.1 | 2 | 3006 | ||||
NR_137176.1 | 2 | 2717 | ||||
NR_137177.1 | 2 | 2696 | ||||
NR_137179.1 | 5 | 2689 | ||||
NR_137180.1 | 5 | 2747 | ||||
NR_137181.1 | 3 | 2632 | ||||
NR_137182.1 | 4 | 2714 | ||||
NR_137183.1 | 3 | 2623 | ||||
NR_137184.1 | 2 | 2541 | ||||
NR_137185.1 | 3 | 2602 | ||||
NR_137186.1 | 3 | 2511 | ||||
NR_137187.1 | 3 | 2490 | ||||
NR_137188.1 | 3 | 2580 | ||||
NR_137189.1 | 2 | 3027 | ||||
NR_137190.1 | 3 | 3060 | ||||
NR_137191.1 | 3 | 3039 | ||||
NR_137192.1 | 4 | 2899 | ||||
NR_137193.1 | 5 | 2726 | ||||
NR_137194.1 | 3 | 2601 | ||||
NR_137195.1 | 2 | 2769 | ||||
NR_137196.1 | 2 | 2790 | ||||
GenBank mRNA | AB075498.1 | - | 2 | 3258 | ||
AK021552.1 | - | 1 | 1903 | |||
AK091713.1 | - | 4 | 1539 | |||
AK123947.1 | - | 2 | 2942 | |||
AL832469.1 | - | 3 | 2294 | |||
BC040599.1 | - | 2 | 2320 | |||
BX538058.1 | - | 2 | 2297 | |||
BX647608.1 | - | 2 | 2298 |