Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST3GAL4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033839.1 | 1 | 129 | |||
GenBank mRNA | AK096370.1 | - | 1 | 129 | ||
BF343672.1 | - | 1 | 129 | |||
DA746418.1 | - | 1 | 129 |