Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102689.1 | 1 | 186 | |||
NR_102691.1 | 1 | 186 | ||||
NR_102692.1 | 1 | 186 | ||||
NR_102693.1 | 1 | 186 | ||||
NR_102694.1 | 1 | 186 | ||||
XM_006720246.3 | XP_006720309.1 | 1 | 114 | |||
GenBank mRNA | BC171914.1 | - | 1 | 145 | ||
BC171918.1 | - | 1 | 145 | |||
GQ871922.1 | - | 1 | 159 | |||
GQ871923.1 | - | 1 | 185 | |||
GQ871926.1 | - | 1 | 97 |
Gene Symbol: DHRS4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198083.3 | NP_932349.2 | 1 | 227 | ||
GenBank mRNA | AK301373.1 | - | 1 | 157 | ||
BC000663.2 | - | 1 | 135 | 3349226 |