Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277864.1 | NP_001264793.1 | 2 | 517 | ||
XM_006720246.3 | XP_006720309.1 | 2 | 209 | |||
GenBank mRNA | GQ871924.1 | - | 2 | 466 |