Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTXN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145668.1 | NP_001139140.1 | 1 | 12 | ||
GenBank mRNA | BX537772.1 | - | 1 | 12 |