Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTXN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145668.1 | NP_001139140.1 | 1 | 144 | ||
XM_017022178.1 | XP_016877667.1 | 2 | 156 | |||
GenBank mRNA | BX537772.1 | - | 1 | 144 |
Gene Symbol: SLC12A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK295375.1 | - | 1 | 49 |