Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL9R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002186.2 | NP_002177.2 | 8 | 1136 | ||
XM_011531151.2 | XP_011529453.1 | 9 | 1186 | |||
XM_011531152.2 | XP_011529454.1 | 9 | 1162 | |||
XM_011531154.2 | XP_011529456.1 | 9 | 1208 | |||
XM_011531155.2 | XP_011529457.1 | 9 | 1195 | |||
XM_011531156.2 | XP_011529458.1 | 9 | 1193 | |||
XM_017029495.1 | XP_016884984.1 | 9 | 1185 | |||
XM_017029496.1 | XP_016884985.1 | 7 | 888 | |||
XM_017029497.1 | XP_016884986.1 | 9 | 1232 | |||
XM_017029498.1 | XP_016884987.1 | 10 | 1337 | |||
XM_017029499.1 | XP_016884988.1 | 10 | 1259 | |||
XM_017029500.1 | XP_016884989.1 | 9 | 1199 | |||
XM_017029502.1 | XP_016884991.1 | 5 | 639 | |||
XM_017029503.1 | XP_016884992.1 | 10 | 1238 | |||
XM_017029504.1 | XP_016884993.1 | 7 | 903 | |||
XM_017029505.1 | XP_016884994.1 | 7 | 885 | |||
XM_017029506.1 | XP_016884995.1 | 2 | 554 | |||
GenBank mRNA | AK303178.1 | - | 9 | 1200 | ||
M84747.1 | - | 8 | 1136 |