Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL9R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002186.2 | NP_002177.2 | 7, 8 | 1070 | ||
XM_011531151.2 | XP_011529453.1 | 8, 9 | 1120 | |||
XM_011531152.2 | XP_011529454.1 | 8, 9 | 1096 | |||
XM_011531154.2 | XP_011529456.1 | 8, 9 | 1142 | |||
XM_011531155.2 | XP_011529457.1 | 8, 9 | 1129 | |||
XM_011531156.2 | XP_011529458.1 | 8, 9 | 1127 | |||
XM_017029495.1 | XP_016884984.1 | 8, 9 | 1119 | |||
XM_017029497.1 | XP_016884986.1 | 8, 9 | 1166 | |||
XM_017029498.1 | XP_016884987.1 | 9, 10 | 1271 | |||
XM_017029499.1 | XP_016884988.1 | 9, 10 | 1193 | |||
XM_017029500.1 | XP_016884989.1 | 8, 9 | 1133 | |||
XM_017029502.1 | XP_016884991.1 | 4, 5 | 573 | |||
XM_017029503.1 | XP_016884992.1 | 9, 10 | 1172 | |||
GenBank mRNA | AK303178.1 | - | 8, 9 | 1134 | ||
M84747.1 | - | 7, 8 | 1070 |