Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024049.1 | 4 | 1190 | |||
NR_024050.1 | 3 | 548 | ||||
NR_131216.1 | 2 | 478 | ||||
NR_131217.1 | 3 | 763 | ||||
NR_131222.1 | 3 | 596 | ||||
GenBank mRNA | BX107175.1 | - | 3 | 519 | ||
HE601934.1 | - | 1 | 332 |