Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207033.1 | NP_001193962.1 | 20 | 5205 | ||
NM_001207034.1 | NP_001193963.1 | 18 | 4908 | |||
NM_001286572.2 | NP_001273501.1 | 20 | 4569 | |||
NM_014771.3 | NP_055586.1 | 20 | 5208 |