Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105549.1 | NP_001099019.1 | 1 | 57 | ||
NM_001105550.1 | NP_001099020.1 | 1 | 57 | |||
NM_001105551.1 | NP_001099021.1 | 1 | 57 | |||
NM_001105552.1 | NP_001099022.1 | 1 | 57 | |||
NM_001277951.1 | NP_001264880.1 | 1 | 57 | |||
NM_001277952.1 | NP_001264881.1 | 1 | 57 | |||
NM_018300.3 | NP_060770.3 | 1 | 57 | |||
GenBank mRNA | AK027518.1 | - | 1 | 51 | ||
BC032867.1 | - | 1 | 18 | 5264072 | ||
BX641726.1 | - | 1 | 17 | |||
DA151844.1 | - | 1 | 14 | |||
DA183557.1 | - | 1 | 57 | |||
DA258790.1 | - | 1 | 12 | |||
DA341245.1 | - | 1 | 29 | |||
DA449506.1 | - | 1 | 22 | |||
DA731895.1 | - | 1 | 51 | |||
DB252431.1 | - | 1 | 31 |