Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256798.1 | NP_001243727.1 | 2 | 556 | ||
XM_005260282.4 | XP_005260339.1 | 2 | 842 | |||
XM_005260283.4 | XP_005260340.1 | 3 | 866 | |||
XM_005260284.4 | XP_005260341.1 | 2 | 141 | |||
XM_005260285.4 | XP_005260342.2 | 2 | 846 | |||
XM_005260286.4 | XP_005260343.1 | 2 | 766 | |||
XM_006723702.3 | XP_006723765.1 | 3 | 233 | |||
XM_006723703.3 | XP_006723766.1 | 2 | 145 | |||
XM_011528562.2 | XP_011526864.1 | 2 | 550 | |||
XM_011528563.1 | XP_011526865.1 | 2 | 550 | |||
XM_017027669.1 | XP_016883158.1 | 2 | 136 | |||
GenBank mRNA | AK097804.1 | - | 4 | 1219 | ||
BQ067609.1 | - | 2 | 311 |