Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021090.3 | NP_066576.1 | 20 | 6826 | ||
NM_153050.2 | NP_694690.1 | 20 | 6742 | |||
NM_153051.2 | NP_694691.1 | 19 | 6715 | |||
XM_005261803.2 | XP_005261860.1 | 22 | 6971 | |||
XM_005261804.3 | XP_005261861.1 | 21 | 6888 | |||
XM_005261805.2 | XP_005261862.1 | 21 | 6944 | |||
XM_005261806.2 | XP_005261863.1 | 21 | 6853 | |||
XM_017029028.1 | XP_016884517.1 | 20 | 6745 | |||
XM_017029029.1 | XP_016884518.1 | 19 | 6708 | |||
XM_017029030.1 | XP_016884519.1 | 19 | 6640 | |||
XM_017029031.1 | XP_016884520.1 | 18 | 6601 | |||
XM_017029033.1 | XP_016884522.1 | 15 | 6172 | |||
XM_017029034.1 | XP_016884523.1 | 21 | 8175 | |||
GenBank mRNA | AK056895.1 | - | 1 | 469 |