Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2E2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136176.1 | 2 | 331 | |||
NR_136177.1 | 2 | 355 | ||||
GenBank mRNA | AI003894.1 | - | 1 | 150 | ||
AI990151.1 | - | 1 | 150 | |||
HY029877.1 | - | 2 | 334 |