Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF732 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137608.1 | NP_001131080.1 | 3 | 1247 | ||
GenBank mRNA | AK302099.1 | - | 3 | 1247 |