Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242729.1 | NP_001229658.1 | 14 | 2466 | ||
XM_006714244.3 | XP_006714307.1 | 12 | 1826 | |||
XM_011532050.2 | XP_011530352.1 | 13 | 2015 | |||
GenBank mRNA | AK125951.1 | - | 5 | 944 |