Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242729.1 | NP_001229658.1 | 9 | 1341 | ||
XM_006714244.3 | XP_006714307.1 | 7 | 701 | |||
XM_011532050.2 | XP_011530352.1 | 8 | 890 | |||
XM_011532051.2 | XP_011530353.1 | 9 | 1356 | |||
GenBank mRNA | DT932593.1 | - | 6 | 652 |