Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019050.2 | NP_061923.2 | 1 | 110 | ||
XM_005263073.3 | XP_005263130.1 | 1 | 137 | |||
XM_005263075.2 | XP_005263132.1 | 1 | 149 | |||
XM_005263076.2 | XP_005263133.1 | 1 | 149 | |||
XM_005263077.3 | XP_005263134.1 | 1 | 149 | |||
XM_006714240.1 | XP_006714303.1 | 1 | 24 | |||
XM_011532038.2 | XP_011530340.1 | 1 | 125 | |||
XM_017008312.1 | XP_016863801.1 | 1 | 154 | |||
XM_017008316.1 | XP_016863805.1 | 1 | 154 | |||
XM_017008317.1 | XP_016863806.1 | 1 | 126 | |||
XM_017008318.1 | XP_016863807.1 | 1 | 126 | |||
GenBank mRNA | BX648836.1 | - | 1 | 152 | ||
CR749490.1 | - | 1 | 102 | |||
DA859085.1 | - | 1 | 95 |