Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019050.2 | NP_061923.2 | 1, 2 | 242 | ||
XM_005263075.2 | XP_005263132.1 | 1, 2 | 281 | |||
XM_005263076.2 | XP_005263133.1 | 1, 2 | 281 | |||
XM_005263077.3 | XP_005263134.1 | 1, 2 | 281 | |||
XM_011532038.2 | XP_011530340.1 | 1, 2 | 257 | |||
XM_017008312.1 | XP_016863801.1 | 1, 2 | 286 | |||
GenBank mRNA | CR749490.1 | - | 1, 2 | 234 | ||
DA886370.1 | - | 1, 2 | 242 |