Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256574.1 | NP_001243503.1 | 1 | 378 | ||
NM_001256575.1 | NP_001243504.1 | 1 | 378 | |||
NM_001256576.1 | NP_001243505.1 | 1 | 378 | |||
NM_003633.3 | NP_003624.1 | 1 | 378 | |||
NR_046318.1 | 1 | 378 | ||||
XM_011543696.2 | XP_011541998.1 | 1 | 378 | |||
XM_011543697.2 | XP_011541999.1 | 1 | 378 | |||
GenBank mRNA | AK096578.1 | - | 1 | 378 |