Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01623 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033379.1 | 2 | 436 | |||
GenBank mRNA | AK055503.1 | - | 2 | 109 | ||
AK291391.1 | - | 2 | 436 |