Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206922.2 | NP_996805.2 | 6 | 463 | ||
XM_005249103.3 | XP_005249160.1 | 6 | 2376 | |||
XM_011514609.2 | XP_011512911.1 | 6 | 2376 | |||
XM_017010856.1 | XP_016866345.1 | 5 | 2244 | |||
XM_017010857.1 | XP_016866346.1 | 4 | 2172 | |||
GenBank mRNA | AY555741.1 | - | 6 | 463 | ||
AY555742.1 | - | 6 | 463 | |||
AY555743.1 | - | 6 | 463 | |||
AY555744.1 | - | 6 | 463 | |||
CN282994.1 | - | 5 | 410 |