Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105377879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BI460215.1 | - | 1, 2 | 284 | ||
CR745838.1 | - | 1, 2 | 167 |