Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM225A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024366.1 | 3 | 3210 | |||
GenBank mRNA | AK125961.1 | - | 1 | 550 | ||
AK127516.1 | - | 1 | 2196 | |||
AL832752.1 | - | 3 | 3033 | |||
CR627046.1 | - | 2 | 3400 | |||
DQ580394.1 | - | 1 | 16 |
Gene Symbol: FAM225B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024376.1 | 3 | 3210 |