Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD99P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033380.1 | 4 | 403 | |||
NR_033381.1 | 4 | 403 | ||||
GenBank mRNA | AK125149.1 | - | 4 | 276 | ||
BC019893.1 | - | 4 | 274 |