Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163941.1 | NP_001157413.1 | 6 | 581 | ||
GenBank mRNA | AB353947.1 | - | 6 | 581 | ||
AY230001.1 | - | 6 | 474 |