Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105375396 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317905.1 | NP_001304834.1 | 10 | 1157 | ||
XM_017011679.1 | XP_016867168.1 | 10 | 879 | |||
GenBank mRNA | KF595048.1 | - | 10 | 1157 |