Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOHLH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012415.2 | NP_001012415.2 | 3, 4 | 395 | ||
NM_001101677.1 | NP_001095147.1 | 3, 4 | 395 | |||
XM_005263403.3 | XP_005263460.1 | 3, 4 | 346 | |||
XM_006717109.3 | XP_006717172.1 | 4, 5 | 1471 | |||
XM_011518698.2 | XP_011517000.1 | 5, 6 | 1500 | |||
GenBank mRNA | AY884306.1 | - | 3, 4 | 340 | ||
AY902244.1 | - | 3, 4 | 395 | |||
BC031861.1 | - | 3, 4 | 406 | 4826967 | ||
EB710265.1 | - | 1, 2 | 92 |