Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001794.3 | NP_001001794.3 | 18 | 1614 | ||
XM_005261914.3 | XP_005261971.1 | 19 | 1789 | |||
XM_011530692.2 | XP_011528994.1 | 19 | 1804 | |||
XM_011530693.2 | XP_011528995.1 | 18 | 1583 | |||
XM_017028801.1 | XP_016884290.1 | 17 | 1625 | |||
GenBank mRNA | AA864612.1 | - | 3 | 412 | ||
AK054743.1 | - | 15 | 2061 | |||
BC031069.1 | - | 18 | 1614 | 5296726 | ||
BM981037.1 | - | 3 | 405 |