Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCQ-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036502.1 | 1 | 315 | |||
NR_036503.1 | 1 | 315 | ||||
GenBank mRNA | AY007155.1 | - | 1 | 201 | ||
CN429636.1 | - | 1 | 175 |