Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT7P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027269.1 | 1 | 195 | |||
GenBank mRNA | BC042387.1 | - | 1 | 195 | ||
BC048143.1 | - | 1 | 195 | |||
BC171738.1 | - | 1 | 161 |
Gene Symbol: LOC100287497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136590.1 | 1 | 229 | |||
NR_136591.1 | 1 | 229 | ||||
NR_136592.1 | 1 | 229 | ||||
NR_136593.1 | 1 | 229 | ||||
GenBank mRNA | DA132394.1 | - | 1 | 212 | ||
DA229030.1 | - | 1 | 212 |
Gene Symbol: LOC645513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037630.1 | 1 | 262 | |||
GenBank mRNA | BM544557.1 | - | 1 | 234 |
Gene Symbol: SEPT7P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024271.1 | 1 | 270 | |||
GenBank mRNA | AL832059.1 | - | 1 | 206 | ||
BC092503.1 | - | 1 | 215 | |||
DA100871.1 | - | 1 | 270 |