Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127711.2 | NP_001121183.1 | 5 | 434 | ||
NM_001286484.1 | NP_001273413.1 | 5 | 459 | |||
NM_030969.4 | NP_112231.3 | 6 | 536 | |||
NR_104454.1 | 4 | 357 | ||||
GenBank mRNA | AK291883.1 | - | 6 | 490 | ||
BC001033.1 | - | 6 | 478 | 3139565 | ||
BC007080.1 | - | 6 | 490 | 4066217 | ||
BC013913.1 | - | 6 | 498 | 3685635 | ||
BC071660.1 | - | 6 | 480 | 4473732 | ||
BG288654.1 | - | 5 | 384 | |||
BP249229.1 | - | 5 | 388 | |||
BU954633.1 | - | 4 | 299 |