Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DKFZp686K1684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_033971.1 | 3 | 617 | |||
GenBank mRNA | BX648962.1 | - | LIT3414 Pax6OS |
3 | 617 |